missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GTF2H2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA565462
This item is not returnable.
View return policy
Description
Immunogen sequence: HVSPPPASSSS ECSLIRMGFP QHTIASLSDQ DAKPSFSMAH LDGNTEPGLT LGGYFCPQCR AKYCELPVEC KICG Highest antigen sequence identity to the following orthologs - mouse 97%, rat 96%.
Initiation of transcription from protein-coding genes in eukaryotes is a complex process that requires RNA polymerase II, as well as families of basal transcription factors. Binding of the factor TFIID (TBP) to the TATA box is believed to be the first step in the formation of a multiprotein complex containing several additional factors, including TFIIA, TFIIB, TFIIE, TFIIF and TFII. TFIIH (or BTF2) is a multisubunit transcription/DNA repair factor that possesses several enzymatic activities. The core of TFIIH is composed of five subunits, designated p89 (XPB or ERCC3), p62, p52, p44 and p34. Additional subunits of the TFIIH complex are p80 (XPD or ERCC2) and the ternary kinase complex composed of Cdk7, cyclin H and MAT1. Both p89 and p80 have ATP-dependent helicase activity. The p62, p52 and p44 subunits have been shown to be involved in nucleotide excision repair.
Specifications
| GTF2H2 | |
| Polyclonal | |
| Unconjugated | |
| Gtf2h2 | |
| 44kDa; basal transcription factor 2, p44 subunit; basic transcription factor 2 44 kDa subunit; BTF2; BTF2 p44; BTF2P44; BTF2-p44; general transcription factor II H, polypeptide 2; general transcription factor II H, polypeptide 2 (44 kDa subunit); general transcription factor IIH polypeptide 2; General transcription factor IIH subunit 2; general transcription factor IIH, polypeptide 2; general transcription factor IIH, polypeptide 2 (44 kDa subunit); general transcription factor IIH, polypeptide 2, 44kD subunit; general transcription factor IIH, polypeptide 2, 44kDa; Gtf2h2; p44; T-BTF2P44; TFIIH; TFIIH basal transcription factor complex p44 subunit | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 2966 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q13888 | |
| Gtf2h2 | |
| Recombinant Human GTF2H2. Recombinant protein control fragment (Product #RP-104829). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction