missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GTF2H2 Mouse anti-Human, Clone: PCRP-GTF2H2-1B9, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-07348-100ug
This item is not returnable.
View return policy
Description
GTF2H2 Monoclonal specifically detects GTF2H2 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Protein Array.
Specifications
| GTF2H2 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 1 to 2 μg/million cells, Immunocytochemistry/Immunofluorescence 1 to 2 μg/mL, Protein Array | |
| Basic transcription factor 2 44 kDa subunit, BTF2, BTF2 p44, BTF2P44TFIIH basal transcription factor complex p44 subunit, General transcription factor IIH polypeptide 2, general transcription factor IIH subunit 2, general transcription factor IIH, polypeptide 2 (44kD subunit), general transcription factor IIH, polypeptide 2, 44kD subunit, general transcription factor IIH, polypeptide 2, 44kDa, MGC102806, T-BTF2P44, TFIIH | |
| Recombinant full-length human GTF2H2 protein | |
| 100 μg | |
| Stem Cell Markers | |
| 2966 | |
| Store at 4°C. | |
| IgG1 |
| Flow Cytometry, Immunofluorescence, Peptide Array | |
| PCRP-GTF2H2-1B9 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction