missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GTF2F2 Recombinant Rabbit Monoclonal Antibody (23GB6490), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£162.00 - £387.00
Specifications
| Antigen | GTF2F2 |
|---|---|
| Clone | 23GB6490 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30088800
![]() |
Invitrogen™
MA553056 |
100 μL |
£387.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30088806
|
Invitrogen™
MA553057 |
20 μL |
£162.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GTF2F2 Recombinant Monoclonal antibody specifically detects GTF2F2 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| GTF2F2 | |
| Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| P13984, Q01750, Q8R0A0 | |
| 2963, 68705, 81674 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB6490 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| 1110031C13Rik; AI326315; ATP-dependent helicase GTF2F2; BTF4; General transcription factor IIF 30 kDa subunit; general transcription factor IIF subunit 2; general transcription factor IIF, polypeptide 2; general transcription factor IIF, polypeptide 2 (30kD subunit); general transcription factor IIF, polypeptide 2, 30kDa; GTF2F2; RAP30; TF2F2; TFIIF; TFIIF beta; TFIIF-beta; transcription factor gamma; transcription initiation factor IIF subunit beta; Transcription initiation factor RAP30; Unknown (protein for MGC:133695) | |
| A synthesized peptide derived from GTF2F2 (1-29AA). | |
| Primary | |
| -20°C | |
| GTF2F2 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title