missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GTF2F1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA554606
This item is not returnable.
View return policy
Description
Immunogen sequence: LSNKKIYQEE EMPESGAGSE FNRKLREEAR RKKYGIVLKE FRPEDQPWLL RVNGKSGRKF KGIKKGGVTE NTSYYIFTQC PDGAFEAFPV HNWYNFTPLA RHRTLTAEEA EEEWERRNKV LNHFSIMQQR RLKDQDQD Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.
TFIIF is a general transcription initiation factor that binds to RNA polymerase II and helps to recruit it to the initiation complex in collaboration with TFIIB. It promotes transcription elongation.
Specifications
| GTF2F1 | |
| Polyclonal | |
| Unconjugated | |
| Gtf2f1 | |
| 2810405L04Rik; BTF4; C76800; General transcription factor IIF 74 kDa subunit; general transcription factor IIF polypeptide 1 (74kD subunit); general transcription factor IIF subunit 1; general transcription factor IIF, polypeptide 1; general transcription factor IIF, polypeptide 1, 74kDa; Gtf2f1; MGC94148; OTTHUMP00000237859; RAP74; TF2F1; TFIIF; TFIIF-alpha; transcription initiation factor IIF subunit alpha; Transcription initiation factor RAP74 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 2962 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P35269 | |
| Gtf2f1 | |
| Recombinant protein corresponding to Human GTF2F1. Recombinant protein control fragment (Product #RP-102509). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction