missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GTF2E2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA528923
This item is not returnable.
View return policy
Description
Recommended positive controls: HeLa, HeLa nucleus. Predicted reactivity: Mouse (92%), Rat (94%), Xenopus laevis (80%), Pig (95%), Chicken (81%), Rhesus Monkey (98%), Bovine (93%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The general transcription factor IIE (TFIIE) is part of the RNA polymerase II transcription initiation complex, recruiting TFIIH and being essential for promoter clearance by RNA polymerase II. TFIIE is a heterodimer (and sometimes heterotetramer) of alpha and beta subunits. The protein encoded by this gene represents the beta subunit of TFIIE.
Specifications
| GTF2E2 | |
| Polyclonal | |
| Unconjugated | |
| GTF2E2 | |
| 34kDa; AI462509; C330006J08Rik; FE; general transcription factor II E, polypeptide 2 (beta subunit); general transcription factor II E, polypeptide 2 (beta subunit, 34kD); general transcription factor IIE subunit 2; general transcription factor IIE, polypeptide 2 (beta subunit); general transcription factor IIE, polypeptide 2 (beta subunit, 34kD); general transcription factor IIE, polypeptide 2, beta; general transcription factor IIE, polypeptide 2, beta 34kDa; Gtf2e2; T2EB; TF2E2; TFIIE beta subunit; TFIIE-B; TFIIE-beta; transcription initiation factor IIE subunit beta; TTD6 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 2961 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1.01 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| P29084 | |
| GTF2E2 | |
| Recombinant protein encompassing a sequence within the center region of human TFIIE beta. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction