missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GTF2E1 Recombinant Rabbit Monoclonal Antibody (23GB6310), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£162.00 - £387.00
Specifications
| Antigen | GTF2E1 |
|---|---|
| Clone | 23GB6310 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30088483
![]() |
Invitrogen™
MA553018 |
100 μL |
£387.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30087815
|
Invitrogen™
MA553019 |
20 μL |
£162.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GTF2E1 Recombinant Monoclonal antibody specifically detects GTF2E1 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Western BlotSpecifications
| GTF2E1 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| P29083, Q9D0D5 | |
| 2960, 303918, 74197 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB6310 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| 2610024P03Rik; 56kDa; AI303800; FE; general transcription factor II E, polypeptide 1 (alpha subunit); general transcription factor II E, polypeptide 1 (alpha subunit, 56kDa); general transcription factor IIE 56 kDa subunit; General transcription factor IIE subunit 1; General transcription factor IIE subunit 1-like protein; general transcription factor IIE, polypeptide 1 (alpha subunit); general transcription factor IIE, polypeptide 1 (alpha subunit, 56kDa); general transcription factor IIE, polypeptide 1, alpha; general transcription factor IIE, polypeptide 1, alpha 56kDa; GTF2E1; TF2E1; TFIIE alpha subunit; TFIIE-A; TFIIE-alpha; Transcription initiation factor IIE subunit alpha | |
| A synthesized peptide derived from human TFIIE alpha (400-439AA). | |
| Primary | |
| -20°C | |
| GTF2E1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title