missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GTF2E1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24405-1-AP-20UL
This item is not returnable.
View return policy
Description
The general transcription factor TFIIE has an essential role in eukaryotic transcription initiation together with RNA polymerase II and other general factors. Human TFIIE consists of two subunits of relative molecular mass 57, 000 (TFIIE-alpha) and 34, 000 (TFIIE-beta) and joins the preinitiation complex after RNA polymerase II and TFIIF. TFIIE-alpha is necessary for transcription initiation together with TFIIE-beta, and recombit TFIIE-alpha can fully replace the natural subunit in an in vitro transcription assay. Its sequence contains several interesting structural motifs (leucine repeat, zinc finger and helix-turn-helix) and sequence similarities to bacterial sigma factors that suggest direct involvement in the regulation of transcription initiation.Specifications
| GTF2E1 | |
| Polyclonal | |
| Unconjugated | |
| GTF2E1 | |
| FE, GTF2E1, TF2E1, TFIIE A, TFIIE alpha | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 2960 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.39 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P29083 | |
| GTF2E1 | |
| GTF2E1 Fusion Protein Ag19794 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction