missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GTF2E1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA566010
This item is not returnable.
View return policy
Description
Immunogen sequence: QSKDHAATTAG AASLAGGHHR EAWATKGPSY EDLYTQNVVI NMDDQEDLHR ASLEGKSAKE RPIWLRESTV QGAYGSEDMK EGGIDMDAFQ EREEGHAGPD DNEEVMRALL IHEKKTSSAM AGSVGAAAPV TAANGSDSE Highest antigen sequence identity to the following orthologs - mouse 88%, rat 88%.
The general transcription factor TFIIE has an essential role in eukaryotic transcription initiation together with RNA polymerase II and other general factors. Human TFIIE consists of two subunits of relative molecular mass 57, 000 (TFIIE-alpha) and 34, 000 (TFIIE-beta) and joins the preinitiation complex after RNA polymerase II and TFIIF. TFIIE-alpha is necessary for transcription initiation together with TFIIE-beta, and recombinant TFIIE-alpha can fully replace the natural subunit in an in vitro transcription assay. Its sequence contains several interesting structural motifs (leucine repeat, zinc finger and helix-turn-helix) and sequence similarities to bacterial sigma factors that suggest direct involvement in the regulation of transcription initiation.
Specifications
| GTF2E1 | |
| Polyclonal | |
| Unconjugated | |
| GTF2E1 | |
| 2610024P03Rik; 56kDa; AI303800; FE; general transcription factor II E, polypeptide 1 (alpha subunit); general transcription factor II E, polypeptide 1 (alpha subunit, 56kDa); general transcription factor IIE 56 kDa subunit; General transcription factor IIE subunit 1; General transcription factor IIE subunit 1-like protein; general transcription factor IIE, polypeptide 1 (alpha subunit); general transcription factor IIE, polypeptide 1 (alpha subunit, 56kDa); general transcription factor IIE, polypeptide 1, alpha; general transcription factor IIE, polypeptide 1, alpha 56kDa; GTF2E1; TF2E1; TFIIE alpha subunit; TFIIE-A; TFIIE-alpha; Transcription initiation factor IIE subunit alpha | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 2960 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P29083 | |
| GTF2E1 | |
| Recombinant Human GTF2E1. Recombinant protein control fragment (Product #RP-105039). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction