missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GTF2B Recombinant Rabbit Monoclonal Antibody (JE38-37)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA556550
This item is not returnable.
View return policy
Description
GTF2B Recombinant Monoclonal Antibody for Western Blot
Ubiquitin is a 76 amino acid highly conserved eukaryotic polypeptide that selectively marks cellular proteins for proteolytic degradation by the 26S proteasome. The process of target selection, covalent attachment and shuttle to the 26S proteasome is a vital means of regulating the concentrations of key regulatory proteins in the cell by limiting their lifespans. Polyubiquitination is a common feature of this modification. Serial steps for modification include the activation of ubiquitin, an ATP-dependent formation of a thioester bond between ubiquitin and the enzyme E1, transfer by transacylation of ubiquitin from E1 to the ubiquitin conjugating enzyme E2, and covalent linkage to the target protein directly by E2 or via E3 ligase enzyme. Deubiquitination enzymes also exist to reverse the marking of protein substrates. Posttranslational tagging by Ub is involved in a multitude of cellular processes, including the cell cycle, cell growth and differentiation, embryogenesis, apoptosis, signal transduction, DNA repair, regulation of transcription and DNA replication, transmembrane transport, stress responses, the immune response, and nervous system functions.
Specifications
| GTF2B | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| P62915, P62916, Q00403 | |
| GTF2B | |
| Recombinant protein within human tfiib aa 217-316/316. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Western Blot | |
| JE38-37 | |
| Unconjugated | |
| GTF2B | |
| general transcription factor 2B L homeolog; general transcription factor IIB; general transcription factor TFIIB; GTF2B; gtf2b.L; gtf2b-a; gtf2b-b; LOW QUALITY PROTEIN: transcription initiation factor IIB; MGC137096 protein; RNA polymerase II alpha initiation factor; RNA polymerase II transcription factor IIB; RP11-82K18.1; S300-II; TF2B; tfiib; Transcription initiation factor IIB; Transcription initiation factor IIB-like protein; wu:fc44c11; XELAEV_18023356mg; XLTFIIB; zgc:66258 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 229906, 2959, 81673 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction