missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
GTF2A2 Polyclonal specifically detects GTF2A2 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
| Antigen | GTF2A2 |
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 0.4 ug/ml, Immunocytochemistry/Immunofluorescence 1-4 ug/ml |
| Formulation | PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide |
| Gene Alias | General transcription factor IIA subunit 2, general transcription factor IIA, 2 (12kD subunit), general transcription factor IIA, 2, 12kDa, HsT18745, TF2A2, TFIIA, TFIIA p12 subunit, TFIIA-12, TFIIA-gamma, TFIIAS, Transcription initiation factor IIA gamma chain, transcription initiation factor IIA subunit 2 |
| Gene Symbols | GTF2A2 |
| Host Species | Rabbit |
| Immunogen | This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:AYQLYRNTTLGNSLQESLDELIQSQQITPQLALQVLLQFDKAINAALAQRVRNRVNFRGSLNTYRFCDNVWTFVLNDVEFREVTELIKVDKVKIVACDGKNTGSNTTE |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?