Learn More
Invitrogen™ GTF2A1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5109795
Description
Immunogen sequence: MANSANTNTV PKLYRSVIED VINDVRDIFL DDGVDEQVLM ELKTLWENKL MQSRAVDGFH SEEQQLLLQV QQQHQPQQQQ HHHHHHHQQA QPQQTVPQQA QTQQVLIPAS QQATAPQVIV PDSKLIQHMN ASNMSAAATA ATLALPAGVT PVQQILTNSG QLLQVVRAAN GAQYIFQPQQ SVVLQQQVIP QMQPGGVQAP VIQQVLAPLP GGISPQTGVI IQPQQILFTG NKTQVIPTTV AAPTPAQAQI TATGQQQPQA QPAQTQAPLV.
Accurate transcription initiation on TATA-containing class II genes involves the ordered assembly of RNA polymerase II (POLR2A) and several general initiation factors. One of these factors is TFIIA, which when purified from HeLa extracts consists of 35-, 19-, and 12-kD subunits.
Specifications
| GTF2A1 | |
| Polyclonal | |
| Unconjugated | |
| GTF2A1 | |
| 6330549H03Rik; AA536742; AA959775; AW060250; general transcription factor 2a, 1; general transcription factor II A, 1; general transcription factor IIA 1; general transcription factor IIA subunit 1; general transcription factor IIA, 1; general transcription factor IIa, 1 (37kD and 19kD subunits); general transcription factor IIA, 1, 19/37kDa; glucose regulated protein, 58kD pseudogene; Gtf2a1; Tf2a; TF2A1; TFIIA; TFIIA alpha, p55, isoform 1; TFIIA alpha/beta; TFIIA alpha/beta subunits; TFIIA large subunit; TFIIA p19 subunit; TFIIA p35 subunit; Tfiia1; TFIIA-42; TfIIAa/b; TFIIAL; Transcription initiation factor IIA alpha chain; Transcription initiation factor IIA beta chain; Transcription initiation factor IIA subunit 1; transcription initiation factor TFIIA 42 kDa subunit | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 2957 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.05 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P52655 | |
| GTF2A1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-270 of human GTF2A1 (NP_056943.1). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.