Learn More
Invitrogen™ GTF2A1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA581901
Description
Immunogen sequence: QQAQTQQVLI PASQQATAPQ VIVPDSKLIQ HMNASNMSAA ATAATLALPA GVTPVQQILT NSGQLLQVVR AANGAQYIFQ PQQSVVLQQQ VIPQMQPGGV QAPVIQQVLA PLPGGISPQT GVIIQPQQIL FTGNKTQVIP TTVAAPTPAQ A Highest antigen sequence indentity to the following orthologs: Rat - 98%, Mouse - 95%.
Accurate transcription initiation on TATA-containing class II genes involves the ordered assembly of RNA polymerase II (POLR2A) and several general initiation factors. One of these factors is TFIIA, which when purified from HeLa extracts consists of 35-, 19-, and 12-kD subunits.
Specifications
| GTF2A1 | |
| Polyclonal | |
| Unconjugated | |
| GTF2A1 | |
| 6330549H03Rik; AA536742; AA959775; AW060250; general transcription factor 2a, 1; general transcription factor II A, 1; general transcription factor IIA 1; general transcription factor IIA subunit 1; general transcription factor IIA, 1; general transcription factor IIa, 1 (37kD and 19kD subunits); general transcription factor IIA, 1, 19/37kDa; glucose regulated protein, 58kD pseudogene; Gtf2a1; Tf2a; TF2A1; TFIIA; TFIIA alpha, p55, isoform 1; TFIIA alpha/beta; TFIIA alpha/beta subunits; TFIIA large subunit; TFIIA p19 subunit; TFIIA p35 subunit; Tfiia1; TFIIA-42; TfIIAa/b; TFIIAL; Transcription initiation factor IIA alpha chain; Transcription initiation factor IIA beta chain; Transcription initiation factor IIA subunit 1; transcription initiation factor TFIIA 42 kDa subunit | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 2957 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ChIP Assay, Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P52655 | |
| GTF2A1 | |
| Recombinant protein corresponding to Human GTF2A1. Recombinant protein control fragment (Product #RP-88980). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.