missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GTF2A1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-46216
This item is not returnable.
View return policy
Description
GTF2A1 Polyclonal specifically detects GTF2A1 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Western Blot (Negative).
Specifications
| GTF2A1 | |
| Polyclonal | |
| Unconjugated | |
| Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
| GTF2A1 | |
| The immunogen for this product maps to a region between residue 100 and 150 of human Transcription Initiation Factor IIA Subunit 1 using the numbering given in entry NP_056943.1 (GeneID 2957). | |
| 100 μL | |
| DNA replication Transcription Translation and Splicing | |
| 2957 | |
| Human, Mouse | |
| IgG |
| Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot | |
| 1.0 mg/mL | |
| Immunohistochemistry 1:10-1:500, Immunoprecipitation 2-5 ug/mg lysate, Immunohistochemistry-Paraffin 1:10-1:500, Western Blot (Negative) | |
| General transcription factor IIA subunit 1, general transcription factor IIA, 1, 19/37kDa, glucose regulated protein, 58kD pseudogene, MGC129969, MGC129970, TF2A1, TFIIA, TFIIA alpha, p55, TFIIA-42, TFIIAL, transcription initiation factor IIA subunit 1, Transcription initiation factor TFIIA 42 kDa subunit | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Based on 100% sequence identity, this antibody is predicted to react with Rat, Bovine, Dog, Horse, Rabbit, Guinea pig_10141, Pig, Panda, Orangutan, Rhesus Monkey, Gorilla, Chimpanzee, White-tuftedear marmoset, Chinese hamster, Naked mole rat, African elephant, and European domestic ferret. | |
| Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction