missing translation for 'onlineSavingsMsg'
Learn More

GTF2A1 Antibody, Novus Biologicals™

Product Code. 18358988 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18358988 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18358988 Supplier Novus Biologicals Supplier No. H00002957D01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

GTF2A1 Polyclonal antibody specifically detects GTF2A1 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen GTF2A1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS (pH 7.4)
Gene Accession No. NP_056943.1
Gene Alias General transcription factor IIA subunit 1, general transcription factor IIA, 1, 19/37kDa, glucose regulated protein, 58kD pseudogene, MGC129969, MGC129970, TF2A1, TFIIA, TFIIA alpha, p55, TFIIA-42, TFIIAL, transcription initiation factor IIA subunit 1, Transcription initiation factor TFIIA 42 kDa subunit
Host Species Rabbit
Immunogen GTF2A1 (NP_056943.1, 1 a.a. - 376 a.a.) full-length human protein. MANSANTNTVPKLYRSVIEDVINDVRDIFLDDGVDEQVLMELKTLWENKLMQSRAVDGFHSEEQQLLLQVQQQHQPQQQQHHHHHHHQQAQPQQTVPQQAQTQQVLIPASQQATAPQVIVPDSKLIQHMNASNMSAAATAATLALPAGVTPVQQILTNSGQLLQVVRAANGAQYIFQPQQSVVLQQQVIPQMQPGGVQAPVIQQVLAPLPGGISPQTGVIIQPQQILFTGNKTQVIPTTVAAPTPAQAQITATGQQQPQAQPAQTQAPLVLQVDGTGDTSSEEDEDEEEDYDDDEEEDKEKDGAEDGQVEEEPLNSEDDVSDEEGQELFDTENVVVCQYDKIHRSKNKWKFHLKDGIMNLNGRDYIFSKAIGDAEW
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 2957
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.