missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GTDC1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5112953
This item is not returnable.
View return policy
Description
GTDC1 Polyclonal Antibody for Western Blot, ELISA
The GTDC1 (Glycosyltransferase-Like Domain Containing 1) gene encodes a protein that is hypothesized to function as a glycosyltransferase. This gene has been implicated in several crucial biological processes, particularly in the nervous system. The GTDC1 cDNA is 2954 base pairs in length, encoding a protein of 458 amino acids, and shows high expression in tissues such as the lung, spleen, testis, and peripheral blood leukocytes. Genetic studies have highlighted its potential role in developmental disorders, such as global developmental delay and delayed speech and language development, linked to microdeletions encompassing the GTDC1 gene. Disruption of GTDC1 expression has been shown to affect neural progenitor cells and neurons, indicating its significance in neural development. Additionally, the gene holds evolutionary conservation, sharing 75% and 37% identity with its mouse and fruit fly counterparts, respectively.
Specifications
| GTDC1 | |
| Polyclonal | |
| Unconjugated | |
| GTDC1 | |
| E330008O22Rik; glycosyltransferase like domain containing 1; glycosyltransferase-like 1; glycosyltransferase-like domain containing 1; glycosyltransferase-like domain-containing protein 1; glycosyltransferase-like protein; Gtdc1; Hmat-Xa; mannosyltransferase candidate; mannosyltransferase-like protein Xa; mat-Xa | |
| Rabbit | |
| Protein G | |
| RUO | |
| 79712 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Western Blot | |
| 2 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q4AE62 | |
| GTDC1 | |
| Recombinant Human Glycosyltransferase-like domain-containing protein 1 protein (1-292AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction