missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GSX2 Recombinant Rabbit Monoclonal Antibody (HL2533)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA556732
This item is not returnable.
View return policy
Description
GSX2 Recombinant Monoclonal Antibody for Western Blot, IHC (P)
Specifications
| GSX2 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| P31316, Q9BZM3 | |
| GSX2 | |
| Recombinant fragment of human GSX2. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Zebrafish | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot | |
| HL2533 | |
| Unconjugated | |
| GSX2 | |
| genetic-screened homeobox 2; genomic screened homeo box 2; genomic screened homeo box 2 protein; GS homeobox 2; GSH2; Gsh-2; Gsx2; Homeobox protein GSH-2 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 14843, 170825, 561076 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction