missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GSX1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5144269
This item is not returnable.
View return policy
Description
GSX1 Polyclonal Antibody for IHC (P), ELISA
GSX1 gene ontology annotations related to this gene include adenohypophysis development; hypothalamus development; neuron fate commitment; spinal cord association neuron differentiation.
Specifications
| GSX1 | |
| Polyclonal | |
| Unconjugated | |
| GSX1 | |
| genomic screened homeo box 1; GS homeo box protein 1; GS homeobox 1; Gsh1; Gsh-1; GSX1; Homeobox protein GSH-1 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 219409 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 | |
| Q9H4S2 | |
| GSX1 | |
| Recombinant Human GS homeobox 1 protein (67-147AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction