missing translation for 'onlineSavingsMsg'
Learn More

GSTZ1 Antibody (044), HRP, Novus Biologicals™

Product Code. 18351881 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18351881 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18351881 Supplier Novus Biologicals Supplier No. NBP290273H

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

GSTZ1 Monoclonal specifically detects GSTZ1 in Human samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GSTZ1
Applications ELISA
Classification Monoclonal
Clone 44
Conjugate HRP
Dilution ELISA
Formulation PBS
Gene Alias EC 2.5.1.18, EC 5.2.1.2, glutathione S-alkyltransferase, glutathione S-aralkyltransferase, Glutathione S-transferase zeta 1, glutathione transferase zeta 1, GSTZ1-1maleylacetoacetate isomerase, MAAIglutathione S-aryltransferase, MAI, maleylacetone isomerase, MGC2029, S-(hydroxyalkyl)glutathione lyase
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human GSTZ1 (NP_665877.1; Met1-Ala216).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline metabolism
Primary or Secondary Primary
Gene ID (Entrez) 2954
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.