missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GSTT4 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15887993
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15887993 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15887993 Supplier Invitrogen™ Supplier No. PA569587

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This target displays homology in the following species: Dog: 79%; Guinea Pig: 79%; Horse: 79%; Human: 93%; Rabbit: 79%; Rat: 79%.

Conjugation of reduced glutathione to a wide number of exogenous and endogenous hydrophobic electrophiles.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GSTT4
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Gstt4
Gene Accession No. A0A1W2PR19
Gene Alias glutathione S-transferase theta pseudogene 1; glutathione S-transferase theta-4; glutathione S-transferase, theta 4; GST class-theta-4; Gstt4; GSTTP1; HS322B1A
Gene Symbols Gstt4
Host Species Rabbit
Immunogen synthetic peptide directed towards the C-terminal region of human GSTT4.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25774
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.