Learn More
Invitrogen™ GSTT2B Monoclonal Antibody (K24025_5C8), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA555649
Description
Sequence of this protein is as follows: SQPSRAVYIF AKKNGIPLEL RTVDLVKGQH KSKEFLQINS LGKLPTLKDG DFILTESSAI LIYLSCKYQT PDHWYPSDLQ ARARVHEYLG WHADCIRGTF GIPLWVQVLG PLIGVQVPEE KVERNRTAMD QALQWLEDKF LGDRPFLAGQ QVTLADLMAL EELMQPVALG YELFEGRPRL AAWRGRVEAF LGAELCQEAH SIILSILEQA AKKTLPTPSP
The protein encoded by this gene, glutathione S-transferase (GST) theta 2B (GSTT2B), is a member of a superfamily of proteins that catalyze the conjugation of reduced glutathione to a variety of electrophilic and hydrophobic compounds. Human GSTs can be divided into five main classes: alpha, mu, pi, theta, and zeta. The theta class includes GSTT1, GSTT2, and GSTT2B. GSTT2 and GSTT2B are nearly identical to each other, and share 55% amino acid identity with GSTT1. All three genes may play a role in human carcinogenesis. The GSTT2B gene is a pseudogene in some populations.
Specifications
| GSTT2B | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| P0CG30 | |
| GSTT2B | |
| Recombinant human GSTT2B (immunogen range is 11-230). | |
| 50 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG2a |
| Western Blot | |
| K24025_5C8 | |
| Unconjugated | |
| GSTT2B | |
| glutathione S-transferase theta 2; glutathione S-transferase theta 2B (gene/pseudogene); glutathione S-transferase theta-2; Glutathione S-transferase theta-2B; GST class-theta-2; GSTT2; GSTT2B; GSTT2P | |
| Mouse | |
| Protein A | |
| RUO | |
| 653689 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.