missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GSTT2B Monoclonal Antibody (K24025_5C8), Invitrogen™

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA556029

 View more versions of this product

Product Code. 30197365

  • £570.00 / 200µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Sequence of this protein is as follows: SQPSRAVYIF AKKNGIPLEL RTVDLVKGQH KSKEFLQINS LGKLPTLKDG DFILTESSAI LIYLSCKYQT PDHWYPSDLQ ARARVHEYLG WHADCIRGTF GIPLWVQVLG PLIGVQVPEE KVERNRTAMD QALQWLEDKF LGDRPFLAGQ QVTLADLMAL EELMQPVALG YELFEGRPRL AAWRGRVEAF LGAELCQEAH SIILSILEQA AKKTLPTPSP

The protein encoded by this gene, glutathione S-transferase (GST) theta 2B (GSTT2B), is a member of a superfamily of proteins that catalyze the conjugation of reduced glutathione to a variety of electrophilic and hydrophobic compounds. Human GSTs can be divided into five main classes: alpha, mu, pi, theta, and zeta. The theta class includes GSTT1, GSTT2, and GSTT2B. GSTT2 and GSTT2B are nearly identical to each other, and share 55% amino acid identity with GSTT1. All three genes may play a role in human carcinogenesis. The GSTT2B gene is a pseudogene in some populations.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

GSTT2B
Monoclonal
1 mg/mL
PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
P0CG30
GSTT2B
Recombinant human GSTT2B (immunogen range is 11-230).
200 μL
Primary
Human, Rat
Antibody
IgG2a
Western Blot
K24025_5C8
Unconjugated
GSTT2B
glutathione S-transferase theta 2; glutathione S-transferase theta 2B (gene/pseudogene); glutathione S-transferase theta-2; Glutathione S-transferase theta-2B; GST class-theta-2; GSTT2; GSTT2B; GSTT2P
Mouse
Protein A
RUO
653689
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.