missing translation for 'onlineSavingsMsg'
Learn More

GSTT2B Antibody, DyLight 488, Novus Biologicals™

Product Code. 18349053 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18349053 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18349053 Supplier Novus Biologicals Supplier No. NBP306612G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

GSTT2B Polyclonal antibody specifically detects GSTT2B in Human samples. It is validated for Western Blot, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen GSTT2B
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate DyLight 488
Formulation 50mM Sodium Borate
Gene Alias glutathione S-transferase theta 2, Glutathione S-Transferase Theta 2B (Gene/Pseudogene), Glutathione S-transferase theta-2B, GST class-theta-2, GSTT2P
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Human GSTT2B (NP_000845.1; Met1-Pro244).
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 653689
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.