missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GSTT2 Antibody (AT10H4), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50577APC
This item is not returnable.
View return policy
Description
GSTT2 Monoclonal specifically detects GSTT2 in Human samples. It is validated for Flow Cytometry.
Specifications
| GSTT2 | |
| Monoclonal | |
| APC | |
| PBS with 0.05% Sodium Azide | |
| GSTT2 | |
| Recombinant human GSTT2 (1-244aa) purified from E. coli | |
| 100 μL | |
| Cancer | |
| 2953 | |
| Store at 4C in the dark. | |
| IgG2b κ |
| Flow Cytometry | |
| AT10H4 | |
| Flow Cytometry | |
| EC 2.5.1.18, glutathione S-transferase theta 2, glutathione S-transferase theta-2, Glutathione S-transferase theta-2B, GST class-theta-2, GSTT2B, MGC182032 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction