missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GSTT1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA522011
This item is not returnable.
View return policy
Description
Recommended positive controls: HepG2, mouse spleen, rat liver. Predicted reactivity: Mouse (81%), Rat (80%), Rhesus Monkey (97%), Bovine (86%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Glutathione S-transferase (GST) theta 1 (GSTT1) is a member of a superfamily of proteins that catalyze the conjugation of reduced glutathione to a variety of electrophilic and hydrophobic compounds. Human GSTs can be divided into five main classes: alpha, mu, pi, theta, and zeta. The theta class includes GSTT1 and GSTT2. The GSTT1 and GSTT2 share 55% amino acid sequence identity and both of them were claimed to have an important role in human carcinogenesis. The GSTT1 gene is located approximately 50kb away from the GSTT2 gene. The GSTT1 and GSTT2 genes have a similar structure, being composed of five exons with identical exon/intron boundaries.
Specifications
| GSTT1 | |
| Polyclonal | |
| Unconjugated | |
| Gstt1 | |
| AI255817; Glutathione S-transferase 1 (theta); glutathione S-transferase 5; glutathione S-transferase theta 1; glutathione S-transferase theta-1; glutathione S-transferase, theta 1; glutathione transferase T1-1; GST 5-5; GST class-theta-1; GSTT1; Gstt1-1; GSTYRS; similar to Glutathione S-transferase theta 1 (GST class-theta 1) (Glutathione transferase T1-1) | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 14871, 25260, 2952 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 1 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| P30711, Q01579, Q64471 | |
| Gstt1 | |
| Full length human GSTT1 Recombinant protein. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction