missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GSTT1 Mouse anti-Human, Clone: 38D11, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59453-50UL
This item is not returnable.
View return policy
Description
GSTT1 Monoclonal specifically detects GSTT1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| GSTT1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500, Flow Cytometry 1:10-1:1056, ELISA, Immunocytochemistry/Immunofluorescence 1:100 | |
| EC 2.5.1.18, glutathione S-transferase theta 1, glutathione S-transferase theta-1, Glutathione transferase T1-1, GST class-theta-1 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
| 38D11 | |
| Unconjugated | |
| Liquid. In PBS (pH 7.4), 10% Glycerol with 0.02% Sodium Azide | |
| GSTT1 | |
| Recombinant human GSTT1 (1-240aa) purified from E. coli | |
| 50 μL | |
| Asthma, Cancer, Immunology | |
| 2952 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction