missing translation for 'onlineSavingsMsg'
Learn More

GSTT1 Mouse anti-Human, Clone: 38D11, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP2-59453-100UL

 View more versions of this product

Product Code. 18246523

  • £378.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

GSTT1 Monoclonal specifically detects GSTT1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

GSTT1
Monoclonal
1 mg/mL
Western Blot 1:500, Flow Cytometry 1:10-1:1056, ELISA, Immunocytochemistry/Immunofluorescence 1:100
EC 2.5.1.18, glutathione S-transferase theta 1, glutathione S-transferase theta-1, Glutathione transferase T1-1, GST class-theta-1
Mouse
Protein A purified
RUO
Primary
Human
Purified
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence
38D11
Unconjugated
Liquid. In PBS (pH 7.4), 10% Glycerol with 0.02% Sodium Azide
GSTT1
Recombinant human GSTT1 (1-240aa) purified from E. coli
100 μL
Asthma, Cancer, Immunology
2952
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
IgG2b κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only