missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GSTT1 Antibody (38D11), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59453CL1
This item is not returnable.
View return policy
Description
GSTT1 Monoclonal antibody specifically detects GSTT1 in Human samples. It is validated for Flow Cytometry,ELISA,Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| GSTT1 | |
| Monoclonal | |
| CoraFluor 1 | |
| EC 2.5.1.18, glutathione S-transferase theta 1, glutathione S-transferase theta-1, Glutathione transferase T1-1, GST class-theta-1 | |
| Recombinant human GSTT1 (1-240aa) purified from E. coli | |
| 0.1 mL | |
| Asthma, Cancer, Immunology | |
| 2952 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2b κ |
| Flow Cytometry, ELISA, Western Blot, Immunocytochemistry/Immunofluorescence | |
| 38D11 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction