missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GSTO2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA596421
This item is not returnable.
View return policy
Description
Immunogen sequence: PHLTKECLVA LRCGRECTNL KAALRQEFSN LEEILEYQNT TFFGGTCISM IDYLLWPWFE RLDVYGILDC VSHTPALRLW ISAMKWDPTV CALLMDKSIF QGFLNLYFQN NPNAFDFGLC; Positive Samples: Rat testis, Mouse pancreas.
The omega class glutathione transferases (GST; EC 2.5.1.18) have poor activity with common GST substrates, but exhibit novel glutathione-dependent thioltransferase, dehydroascorbate reductase, and monomethylarsonate reductase activities, and they modulate Ca(2+) release by ryanodine receptors (e.g., RYR1).
Specifications
| GSTO2 | |
| Polyclonal | |
| Unconjugated | |
| Gsto2 | |
| 1700020F09Rik; 4930425C18Rik; bA127L20.1; bA127L20.1 (novel glutathione-S-transferase); glutathione S-transferase omega 2; glutathione S-transferase omega 2-2; Glutathione S-transferase omega-2; glutathione-dependent dehydroascorbate reductase; glutathione-S-transferase-like protein; GSTO 2-2; Gsto2; GSTO-2; LOC100720364; MMA(V) reductase; monomethylarsonic acid reductase | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 309465, 68214 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.91 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q6AXV9, Q8K2Q2 | |
| Gsto2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 124-243 of human GSTO2 (NP_8990621). | |
| 100 μL | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction