missing translation for 'onlineSavingsMsg'
Learn More

GSTO2 Antibody, Novus Biologicals™

Product Code. 18407371 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18407371 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18407371 Supplier Novus Biologicals Supplier No. NBP214075

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

GSTO2 Polyclonal antibody specifically detects GSTO2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen GSTO2
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.04 to 0.4 μg/mL, Immunohistochemistry 1:50 to 1:200, Immunocytochemistry/ Immunofluorescence 0.25 to 2 μg/mL, Immunohistochemistry-Paraffin 1:50 to 1:200
Formulation PBS (pH 7.2) and 40% Glycerol
Gene Alias bA127L20.1, bA127L20.1 (novel glutathione-S-transferase), EC 2.5.1.18, glutathione S-transferase omega 2, glutathione S-transferase omega-2, glutathione-S-transferase-like protein, GSTO-2
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to the amino acids: DVYGILDCVSHTPALRLWISAMKWDPTVCALLMDKSIFQGFLNLYFQNNPNAFDFG
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline metabolism
Primary or Secondary Primary
Gene ID (Entrez) 119391
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.