missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GSTO-IN-2, MedChemExpress
Description
GSTO-IN-2 is a glutathione S-transferase inhibitor with IC50s of 3.6, 16.3, and 1.4 μM for GSTA2, GSTM1, and GSTP1-1.
Specifications
Specifications
| Chemical Name or Material | GSTO-IN-2 |
| Physical Form | Solid |
| CAS | 1202710-57-3 |
| Color | White |
| Quantity | 5 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C33H52N2O9 |
| For Use With (Application) | Neuroscience-Neuromodulation |
| Solubility Information | DMSO : 100 mg/mL (161.09 mM; Need ultrasonic) |
| SMILES | C[C@@]1(-;@[C@@]2([H])[C@H](C)CCC(O)=O)-;@[C@](-;@C-;@C-;@2)([H])-;@[C@@](-;@C-;@C-;@[C@@]3([H])-;@[C@@]-;@4(-;@C-;@C-;@[C@@H](OC([C@@H](NC(CC[C@@H](C(O)=O)N)=O)CC(O)=O)=O)-;@C-;@3)C)([H])-;@[C@]4([H])-;@C-;@C-;@1 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?