missing translation for 'onlineSavingsMsg'
Learn More

GSTM4 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Product Code. 18370734 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18370734 100 μL 100µL
18331007 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18370734 Supplier Novus Biologicals Supplier No. NBP304533100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

GSTM4 Polyclonal specifically detects GSTM4 in Human, Mouse samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GSTM4
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias EC 2.5.1.18, glutathione S-alkyltransferase M4, glutathione S-aralkyltransferase M4, glutathione S-aryltransferase M4, glutathione S-transferase M4, glutathione S-transferase mu 4, GST class-mu 4, GSTM4-4, GST-Mu2, GTM4, GTS-Mu2, MGC131945, MGC9247, S-(hydroxyalkyl)glutathione lyase M4
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-218 of human GSTM4 (NP_000841.1).
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2948
Target Species Human, Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.