missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
GSTM2 Polyclonal specifically detects GSTM2 in Human samples. It is validated for Western Blot, ELISA.
Specifications
Specifications
| Antigen | GSTM2 |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 700 |
| Dilution | Western Blot, ELISA |
| Formulation | 50mM Sodium Borate |
| Gene Alias | glutathione S-transferase mu 2 (muscle), MGC117303, muscle, S-(hydroxyalkyl)glutathione lyase M2 |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with purified, recombinant GSTM2 (P28161; Met 1-Lys 218). |
| Purification Method | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?