missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GSTK1 Recombinant Rabbit Monoclonal Antibody (JE47-38)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA534852
This item is not returnable.
View return policy
Description
Positive Control: SK-BR-3, human skin tissue, human placenta tissue, human liver cancer tissue, human kidney tissue, human pancreas tissue, SiHa.
GSTK1 encodes a member of the kappa class of the glutathione transferase superfamily of enzymes that function in cellular detoxification. The encoded protein is localized to the peroxisome and catalyzes the conjugation of glutathione to a wide range of hydrophobic substates facilitating the removal of these compounds from cells. Alternative splicing results in multiple transcript variants.
Specifications
| GSTK1 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q9Y2Q3 | |
| GSTK1 | |
| Recombinant protein within Human GSTK1 aa 1-140. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| JE47-38 | |
| Unconjugated | |
| GSTK1 | |
| 0610025I19Rik; AW260476; DsbA-L; glutathione S-transferase class kappa; glutathione S-transferase k1; glutathione S-transferase kappa 1; Glutathione S-transferase subunit 13; glutathione S-transferase subunit 13 homolog; glutathione S-transferase, mitochondrial; GST; GST 13-13; GST class-kappa; GST13; GST13-13; Gstk1; GSTK1-1; GSTkappa; HDCMD47P; hGSTK1; mGSTK1; rGSTK1 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 373156 | |
| -20°C, Avoid Freeze/Thaw Cycles, store in dark | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction