missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GSTK1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA535381
This item is not returnable.
View return policy
Description
This antibody has been tested in 3T3-L1 adipocytes and mouse white and brown adipose tissue lysate samples. It contains enough material to conduct 10 mini-Western blots. It is recommended to add 1% BSA to the antibody dilution buffer.
GSTK1 encodes a member of the kappa class of the glutathione transferase superfamily of enzymes that function in cellular detoxification. The encoded protein is localized to the peroxisome and catalyzes the conjugation of glutathione to a wide range of hydrophobic substates facilitating the removal of these compounds from cells. Alternative splicing results in multiple transcript variants.
Specifications
| GSTK1 | |
| Polyclonal | |
| Unconjugated | |
| GSTK1 | |
| 0610025I19Rik; AW260476; DsbA-L; glutathione S-transferase class kappa; glutathione S-transferase k1; glutathione S-transferase kappa 1; Glutathione S-transferase subunit 13; glutathione S-transferase subunit 13 homolog; glutathione S-transferase, mitochondrial; GST; GST 13-13; GST class-kappa; GST13; GST13-13; Gstk1; GSTK1-1; GSTkappa; HDCMD47P; hGSTK1; mGSTK1; rGSTK1 | |
| Rabbit | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| Immunohistochemistry, Western Blot | |
| Conc. Not Determined | |
| whole serum with 40% glycerol and no preservative | |
| Q9DCM2, Q9Y2Q3 | |
| GSTK1 | |
| Full length recombinant mouse DsbA-L protein. | |
| RUO | |
| 373156, 76263 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction