missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GSTA5 Antibody, Novus Biologicals™
Mouse Polyclonal Antibody
Brand: Novus Biologicals H00221357-B01P
This item is not returnable.
View return policy
Description
GSTA5 Polyclonal antibody specifically detects GSTA5 in Human samples. It is validated for Western Blot
Specifications
| GSTA5 | |
| Polyclonal | |
| PBS (pH 7.4) | |
| EC 2.5.1.18, glutathione S-transferase A5, Glutathione S-transferase A5-5, glutathione S-transferase alpha 5, glutathione transferase A5, GST class-alpha member 5 | |
| GSTA5 (AAI48817.1, 1 a.a. - 222 a.a.) full-length human protein. MAEKPKLHYSNARGSMESIRWLLAAAGVELEEKFLESAEDLDKLRNDGSLLFQQVPMVEIDGMKLVQTRAILNYIASKYNLYGKDMKERALIDMYTEGIVDLTEMILLLLICQPEERDAKTALVKEKIKNRYFPAFEKVLKSHRQDYLVGNKLSWADIHLVELFYYVEELDSSLISSFPLLKALKTRISNLPTVKKFLQPGSQRKPPMDEKSLEEARKIFRF | |
| 0.05 mg | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| AAI48817.1 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 221357 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction