missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GSTA4 Antibody (OTI3F5), Alexa Fluor™ 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70856AF405
This item is not returnable.
View return policy
Description
GSTA4 Monoclonal specifically detects GSTA4 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.
Specifications
| GSTA4 | |
| Monoclonal | |
| Alexa Fluor 405 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| GSTA4 | |
| Full length recombinant protein of human GSTA4 (NP_001503) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| OTI3F5 | |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| DKFZp686D21185, EC 2.5.1.18, glutathione S-alkyltransferase A4, glutathione S-aralkyltransferase A4, glutathione S-aryltransferase A4, glutathione S-transferase A4, Glutathione S-transferase A4-4, glutathione S-transferase alpha 4, glutathione transferase A4-4, GST class-alpha member 4, GSTA4-4, GTA4, S-(hydroxyalkyl)glutathione lyase A4 | |
| Mouse | |
| Affinity Purified | |
| metabolism | |
| 2941.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction