Learn More
Invitrogen™ GSTA2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA596757
Description
Immunogen sequence: MAEKPKLHYS NIRGRMESIR WLLAAAGVEF EEKFIKSAED LDKLRNDGYL MFQQVPMVEI DGMKLVQTRA ILNYIASKYN LYGKDIKEKA LIDMYIEGIA DLGEMILLLP FSQPEEQDAK LALIQEKTKN RYFPAFEKVL KSHGQDYLVG NKLSRADIHL VELLYYVEEL DSSLISSFPL LKALKTRISN LPTVKKFLQP GSPRKPPMDE KSLEESRKIF RF; Positive Samples: Mouse liver, Mouse testis, Rat liver, Rat kidney; Cellular Location: Cytoplasm.
Cytosolic and membrane-bound forms of glutathione S-transferase are encoded by two distinct supergene families. These enzymes function in the detoxification of electrophilic compounds, including carcinogens, therapeutic drugs, environmental toxins and products of oxidative stress, by conjugation with glutathione. The genes encoding these enzymes are known to be highly polymorphic. These genetic variations can change an individual's susceptibility to carcinogens and toxins as well as affect the toxicity and efficacy of some drugs. At present, eight distinct classes of the soluble cytoplasmic mammalian glutathione S-transferases have been identified: alpha, kappa, mu, omega, pi, sigma, theta and zeta. This gene encodes a glutathione S-tranferase belonging to the alpha class. The alpha class genes, located in a cluster mapped to chromosome 6, are the most abundantly expressed glutathione S-transferases in liver. In addition to metabolizing bilirubin and certain anti-cancer drugs in the liver, the alpha class of these enzymes exhibit glutathione peroxidase activity thereby protecting the cells from reactive oxygen species and the products of peroxidation.
Specifications
| GSTA2 | |
| Polyclonal | |
| Unconjugated | |
| Gsta2 | |
| 13-hydroperoxyoctadecadienoate peroxidase; Androst-5-ene-3,17-dione isomerase; glutathione S-alkyltransferase A2; glutathione S-aralkyltransferase A2; glutathione S-aryltransferase A2; glutathione S-transferase A2; glutathione S-transferase alpha 2; glutathione S-transferase alpha-1; Glutathione S-transferase GT41A; glutathione S-transferase Ya subunit; glutathione S-transferase Ya-1; glutathione S-transferase, alpha 2 (Yc2); Glutathione-S-transferase alpha type (Yc2); glutathione-S-transferase, alpha type2; GST 1-1; GST 1a-1a; GST A1-1; GST B; GST class-alpha member 2; GST HA subunit 2; GST Ya1; GST2; Gst2-2; Gsta1; Gsta2; GSTA2-2; Gstc2; Gstc-2; GST-gamma; GTA2; GTH2; Ligandin; liver GST2; S-(hydroxyalkyl)glutathione lyase A2; testis tissue sperm-binding protein Li 59n | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 14858, 24422, 2939 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.24 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P04903, P09210, P10648 | |
| Gsta2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-222 of human GSTA2 (NP_0008373). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.