missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva GST Vector Primers for Sequencing
Shop All Cytiva Products
Click to view available options
Primer:
pGEX 3′ Sequencing
pGEX 5′ Sequencing
Unit Size:
Each
Description
- Provided ready for immediate use
- pGEX 5’ Sequencing Primer: 5’-d[GGGCTGGCAAGCCACGTTTGGTG]-3’
- pGEX 3' Sequencing Primer: 5’-d[CCGGGAGCTGCATGTGTCAGAGG]-3’
Specifications
Specifications
| For Use With (Equipment) | Glutathione S-transferase (GST) gene fusion system |
| Content And Storage | -20°C |
| Primer | pGEX 5′ Sequencing |
| Quantity | 260 U |
| For Use With (Application) | Ready-to-use in sequencing double-stranded DNA inserted into pGEX Vectors |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction