missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ LanthaScreen™ Tb-anti-GST (Rabbit) Antibody
Rabbit Polyclonal Antibody
Brand: Thermo Scientific™ A15114
This item is not returnable.
View return policy
Description
- LanthaScreen™ terbium-labeled anti-GST Rabbit antibodies are used to indirectly label proteins tagged with GST
- This can be advantageous when working with partially purified protein, or in cases in which direct labeling of the protein using amine-reactive Tb chelates negatively impacts protein activity
Specifications
| Kinase Assay | |
| Tb (Terbium)/Lanthanide Chelates | |
| 1 mg | |
| Frozen | |
| Rb | |
| GST | |
| LanthaScreen™ |
| Polyclonal | |
| Rabbit | |
| Terbium-labeled anti-GST antibody in HEPES buffered saline. Store at -20°C. | |
| Microplate Reader | |
| Drug Development | |
| KA |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction