missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ LanthaScreen™ Tb-anti-GST (Rabbit) Antibody
Rabbit Polyclonal Antibody
Brand: Thermo Scientific™ A15112
This item is not returnable.
View return policy
Description
- LanthaScreen™ terbium-labeled anti-GST Rabbit antibodies are used to indirectly label proteins tagged with GST
- This can be advantageous when working with partially purified protein, or in cases in which direct labeling of the protein using amine-reactive Tb chelates negatively impacts protein activity
Specifications
| GST | |
| Polyclonal | |
| in HEPES buffered Saline | |
| 25 μg | |
| Drug Development | |
| Terbium-labeled anti-GST antibody in HEPES buffered saline. Store at -20°C. | |
| Microplate Reader | |
| Drug Development | |
| KA |
| Kinase Assay | |
| Tb (Terbium) | |
| Rabbit | |
| RUO | |
| Primary | |
| Frozen | |
| Rb | |
| GST | |
| LanthaScreen |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
For Research Use Only. Not for use in diagnostic procedures.
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido