missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GST Omega 1 140A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA531185
This item is not returnable.
View return policy
Description
Recommended positive controls: 293T, HepG2, mouse colon, rat liver. Predicted reactivity: Pig (82%), Rhesus Monkey (97%), Chimpanzee (98%), Bovine (84%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene is an omega class glutathione S-transferase (GST) with glutathione-dependent thiol transferase and dehydroascorbate reductase activities. GSTs are involved in the metabolism of xenobiotics and carcinogens. The encoded protein acts as a homodimer and is found in the cytoplasm. Three transcript variants encoding different isoforms have been found for this gene.
Specifications
| GST Omega 1 140A | |
| Polyclonal | |
| Unconjugated | |
| Gsto1 | |
| AA407097; AI194287; AU018802; epididymis secretory protein Li 21; glutathione S-transferase like; glutathione S-transferase omega 1; glutathione S-transferase omega 1-1; Glutathione S-transferase omega-1; Glutathione transferase omega 1 (GSTO 1-1) (p28); glutathione-dependent dehydroascorbate reductase; glutathione-S-transferase like; GSTO 1-1; Gsto1; GSTO-1; GSTTLP28; GSTX; Gtsttl; HEL-S-21; MMA(V) reductase; Monomethylarsonic acid reductase; P28; RP11-99N20.1; S-(Phenacyl)glutathione reductase; SPG-R | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 114846, 14873, 9446 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
| O09131, P78417, Q9Z339 | |
| Gsto1 | |
| Recombinant protein encompassing a sequence within the center region of human GSTO1. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction