missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GSS Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA562976
This item is not returnable.
View return policy
Description
Immunogen sequence: NLYGEEMVQA LKQLKDSEER ASYILMEKIE PEPFENCLLR PGSPARVVQC ISELGIFGVY VRQEKTLVMN KHVGHLLRTK AI Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 91%.
Glutathione is important for a variety of biological functions, including protection of cells from oxidative damage by free radicals, detoxification of xenobiotics, and membrane transport. The protein encoded by this gene functions as a homodimer to catalyze the second step of glutathione biosynthesis, which is the ATP-dependent conversion of gamma-L-glutamyl-L-cysteine to glutathione. Defects in this gene are a cause of glutathione synthetase deficiency.
Specifications
| GSS | |
| Polyclonal | |
| Unconjugated | |
| GSS | |
| AI314904; Glutathione synthase; glutathione synthetase; GS-A/GS-B; GSH S; GSH synthetase; GSHB; GSHS; GSH-S; GSS; HEL-S-64p; HEL-S-88n; MGC14098; OTTHUMP00000030711 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 2937 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P48637 | |
| GSS | |
| Recombinant protein corresponding to Human GSS. Recombinant protein control fragment (Product #RP-99312). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction