missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GSR Recombinant Rabbit Monoclonal Antibody (23GB2845), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA553479
This item is not returnable.
View return policy
Description
This gene encodes a member of the class-I pyridine nucleotide-disulfide oxidoreductase family. This enzyme is a homodimeric flavoprotein. It is a central enzyme of cellular antioxidant defense, and reduces oxidized glutathione disulfide (GSSG) to the sulfhydryl form GSH, which is an important cellular antioxidant. Rare mutations in this gene result in hereditary glutathione reductase deficiency. Multiple alternatively spliced transcript variants encoding different isoforms have been found.
Specifications
| GSR | |
| Recombinant Monoclonal | |
| 0.125 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P00390, P47791 | |
| Gsr | |
| A synthesized peptide derived from human GLUR (470-522AA). | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB2845 | |
| Unconjugated | |
| Gsr | |
| AI325518; D8Ertd238e; epididymis luminal protein 75; epididymis secretory sperm binding protein Li 122m; GLUR; glutathione reductase; glutathione reductase 1; glutathione reductase, mitochondrial; glutathione S-reductase; glutathione-disulfide reductase; GR; Gr1; Gr-1; GRase; GRD1; GSR; HEL-75; HEL-S-122m; hypothetical protein LOC553575; zgc:110010 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 14782, 2936 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction