missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GSPT2 Antibody (OTI2G9), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71768PE
This item is not returnable.
View return policy
Description
GSPT2 Monoclonal specifically detects GSPT2 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.
Specifications
| GSPT2 | |
| Monoclonal | |
| PE | |
| PBS with 0.05% Sodium Azide | |
| GSPT2 | |
| Full length recombinant protein of human GSPT2 (NP_060564) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Flow Cytometry | |
| OTI2G9 | |
| Flow Cytometry | |
| eRF3b, ERF3BGST2, eukaryotic peptide chain release factor GTP-binding subunit ERF3B, Eukaryotic peptide chain release factor subunit 3b, FLJ10441, G1 to S phase transition 2, G1 to S phase transition protein 2 homolog | |
| Mouse | |
| Affinity Purified | |
| Core ESC Like Genes, Stem Cell Markers | |
| 23708.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction