missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GSNOR/ADH5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11051-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the alcohol dehydrogenase family. Members of this family metabolize a wide variety of substrates, including ethanol, retinol, other aliphatic alcohols, hydroxysteroids, and lipid peroxidation products. The encoded protein forms a homodimer. It has virtually no activity for ethanol oxidation, but exhibits high activity for oxidation of long-chain primary alcohols and for oxidation of S-hydroxymethyl-glutathione, a spontaneous adduct between formaldehyde and glutathione. This enzyme is an important component of cellular metabolism for the elimination of formaldehyde, a potent irritant and sensitizing agent that causes lacrymation, rhinitis, pharyngitis, and contact dermatitis. The human genome contains several non-transcribed pseudogenes related to this gene.Specifications
| GSNOR/ADH5 | |
| Polyclonal | |
| Unconjugated | |
| ADH5 | |
| ADH 3, ADH5, ADHX, Alcohol dehydrogenase 5, Alcohol dehydrogenase class 3, FALDH, FDH, GSH FDH, GSNOR | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 100145871, 11532, 128 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence, Immunocytochemistry | |
| 0.14 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P11766, P12711, P28474 | |
| Adh5 | |
| GSNOR,ADH5 Fusion Protein Ag1533 | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction