missing translation for 'onlineSavingsMsg'
Learn More

GSK840, MedChemExpress

Product Code. p-200096488
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
This item is not returnable. View return policy

Product Code. 16463928

Brand: Medchem Express HY104021

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress GSK840 (GSK'840) is a receptor-interacting protein kinase 3 (RIP3 or RIPK3) inhibitor, which binds RIP3 kinase domain with an IC50 of 0.9 nM, and inhibits kinase activity with an IC50 of 0.3 nM.

GSK840 (GSK'840) is a receptor-interacting protein kinase 3 (RIP3 or RIPK3) inhibitor, which binds RIP3 kinase domain with an IC50 of 0.9 nM, and inhibits kinase activity with an IC50 of 0.3 nM.

Specifications

Chemical Name or Material GSK840
CAS 2361146-30-5
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C21H23N3O3
Quantity 10 mg
Synonym GSK'840
Solubility Information DMSO : 110 mg/mL (301.02 mM; Need ultrasonic)
SMILES O=C(OC(C)(C)C)CC1=CC=C(N2C3=CC=C(C(NC)=O)C=C3N=C2)C=C1
Molecular Weight (g/mol) 365.43
Formula Weight 365.43
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.