missing translation for 'onlineSavingsMsg'
Learn More

GSK840, MedChemExpress

Product Code. 16463928
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16463928 10 mg 10mg
16446647 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16463928 Supplier Medchem Express Supplier No. HY104021
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress GSK840 (GSK'840) is a receptor-interacting protein kinase 3 (RIP3 or RIPK3) inhibitor, which binds RIP3 kinase domain with an IC50 of 0.9 nM, and inhibits kinase activity with an IC50 of 0.3 nM.

GSK840 (GSK'840) is a receptor-interacting protein kinase 3 (RIP3 or RIPK3) inhibitor, which binds RIP3 kinase domain with an IC50 of 0.9 nM, and inhibits kinase activity with an IC50 of 0.3 nM.

Specifications

Chemical Name or Material GSK840
CAS 2361146-30-5
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C21H23N3O3
Quantity 10 mg
Synonym GSK'840
Solubility Information DMSO : 110 mg/mL (301.02 mM; Need ultrasonic)
SMILES O=C(OC(C)(C)C)CC1=CC=C(N2C3=CC=C(C(NC)=O)C=C3N=C2)C=C1
Molecular Weight (g/mol) 365.43
Formula Weight 365.43
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.