missing translation for 'onlineSavingsMsg'
Learn More

GSK591, MedChemExpress

Product Code. 16494266
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16494266 100 mg 100mg
16472546 25 mg 25mg
16459516 50 mg 50mg
16445607 5 mg 5mg
16422868 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16494266 Supplier Medchem Express Supplier No. HY100235
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress GSK591 (EPZ015866) is a potent and selective inhibitor of protein methyltransferase 5 (PRMT5) with an IC50 of 4 nM.

GSK591 (EPZ015866) is a potent and selective inhibitor of protein methyltransferase 5 (PRMT5) with an IC50 of 4 nM.

Specifications

Chemical Name or Material GSK591
CAS 1616391-87-7
Color White
Purity Grade Notes Research
Molecular Formula C22H28N4O2
Quantity 100 mg
Synonym EPZ015866 GSK3203591
Solubility Information DMSO : ≥ 135 mg/mL (354.81 mM)
SMILES O=C(C1=CC(NC2CCC2)=NC=C1)NC[C@H](O)CN3CC4=C(C=CC=C4)CC3
Molecular Weight (g/mol) 380.48
Formula Weight 380.48
Percent Purity 98.86%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.