missing translation for 'onlineSavingsMsg'
Learn More

GSK503, MedChemExpress

Product Code. 16481587
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16481587 50 mg 50mg
16478117 5 mg 5mg
16425408 10 mg 10mg
16466006 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16481587 Supplier Medchem Express Supplier No. HY12856
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress GSK503 is a potent and specific inhibitor of EZH2 methyltransferase with Kiapp values of 3 to 27 nM.

GSK503 is a potent and specific inhibitor of EZH2 methyltransferase with Kiapp values of 3 to 27 nM.

Specifications

Chemical Name or Material GSK503
CAS 1346572-63-1
Color Light Gray
Purity Grade Notes Research
Molecular Formula C31H38N6O2
Quantity 50 mg
Solubility Information DMSO : ≥ 44 mg/mL (83.54 mM)
SMILES O=C(C1=CC(C2=CC=C(N3CCN(C)CC3)N=C2)=CC4=C1C(C)=CN4C(C)C)NCC5=C(C)C=C(C)NC5=O
Molecular Weight (g/mol) 526.67
Formula Weight 526.67
Percent Purity 99.73%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.