missing translation for 'onlineSavingsMsg'
Learn More

GSK484 hydrochloride, MedChemExpress

Product Code. 16405797
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16405797

Brand: Medchem Express HY1005145mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress GSK484 hydrochloride is a selective and reversible peptidylarginine deiminase 4 (PAD4) inhibitor. GSK484 hydrochloride demonstrates high affinity binding to PAD4 with IC50s of 50 nM in the absence of Calcium. In the presence of 2 mM Calcium, notably lower potency (250 nM) is observed.

GSK484 hydrochloride is a selective and reversible peptidylarginine deiminase 4 (PAD4) inhibitor. GSK484 hydrochloride demonstrates high affinity binding to PAD4 with IC50s of 50 nM in the absence of Calcium. In the presence of 2 mM Calcium, notably lower potency (250 nM) is observed.

Specifications

Chemical Name or Material GSK484 hydrochloride
CAS 1652591-81-5
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C27H32ClN5O3
Quantity 5 mg
Solubility Information DMSO : 125 mg/mL (245.08 mM; Need ultrasonic) ∣H2O : 50 mg/mL (98.03 mM; Need ultrasonic)
SMILES O=C(N1C[C@H](N)[C@H](O)CC1)C2=CC(OC)=C3C(N=C(C(N4CC5CC5)=CC6=C4C=CC=C6)N3C)=C2.[H]Cl
Molecular Weight (g/mol) 510.03
Formula Weight 510.03
Percent Purity 97.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.